Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(9-Oxoxanthen-2-yl)propionic Acid 1,8-Diazabicyclo[5.4.0]undec-7-ene Salt 98.0+%, TCI America™
SDP

Catalog No. O04481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O04481G Supplier TCI America Supplier No. O04481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1346753-05-6
Molecular Formula C25H28N2O4
Molecular Weight (g/mol) 420.509
InChI Key ZLFMLTKPDRGWFB-UHFFFAOYSA-N
PubChem CID 134128890
IUPAC Name 2,3,4,6,7,8,9,10-octahydropyrimido[1,2-a]azepine;2-(9-oxoxanthen-2-yl)propanoic acid
SMILES CC(C1=CC2=C(C=C1)OC3=CC=CC=C3C2=O)C(=O)O.C1CCC2=NCCCN2CC1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 420.51
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9-Oxoxanthen-2-yl)propionic Acid 1,8-Diazabicyclo[5.4.0]undec-7-ene Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.