accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13472-56-5

IUPAC Name: 2-methoxy-5-methylpyridine
Synonyms: 2-methoxy-5-methylpyridine
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: NFQGQMBFMIIIOR-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results