accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13472-61-2

IUPAC Name: 5-iodo-2-methoxypyridine
Synonyms: 5-iodo-2-methoxypyridine
Molecular Weight (g/mol): 235.02
Molecular Formula: C6H6INO
InChi Key: NTXRNCUPGYOZCN-UHFFFAOYSA-N
SMILES: COC1=NC=C(I)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)