accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1347815-35-3

IUPAC Name: 2-(pyridin-3-yl)-1H-imidazole-4,5-dicarbothioamide
Synonyms: 2-(pyridin-3-yl)-1H-imidazole-4,5-dicarbothioamide
Molecular Weight (g/mol): 263.34
Molecular Formula: C10H9N5S2
InChi Key: MSRZAVQGAJGQJT-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(N=C(N1)C1=CN=CC=C1)C(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results