accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13494-90-1

IUPAC Name: gallium(3+) trinitrate
Synonyms: gallium nitrate ganite
Molecular Weight (g/mol): 255.74
Molecular Formula: GaN3O9
InChi Key: CHPZKNULDCNCBW-UHFFFAOYSA-N
SMILES: [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results