accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13499-05-3

IUPAC Name: hafnium(4+) tetrachloride
Synonyms: hafnium tetrachloride
Molecular Weight (g/mol): 320.29
Molecular Formula: Cl4Hf
InChi Key: PDPJQWYGJJBYLF-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Hf+4]
Molecular Weight (g/mol) 
  • (9)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (9)
  • (9)
  • (4)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (9)
  • (2)
  • (2)
  • (2)