Learn More
Hafnium(IV) chloride, 99.9% (metals basis), Zr<0.5%
Description
Hafnium(IV) chloride is used as a precursor for catalyst involved in the Ziegler-Natta polymerization of propylene. It is an effective Lewis acid for various applications in organic synthesis viz alkylation of ferrocene. Further, it increases the rate and control of 1,3-dipolar cycloadditions. It is also used to prepare hafnium dioxide and hafnium silicate, which is used as high-k-dielectrics in the production of integrated circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 320° (sublimation) |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Odor | Irritating |
| Sensitivity | Moisture sensitive |
| Assay Percent Range | 99.9% (Metals basis) |
| Solubility Information | Soluble in methanol and acetone. |
| Formula Weight | 320.3 |
| Chemical Name or Material | Hafnium(IV) chloride |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: III |
| EINECSNumber | 236-826-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.