accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHOHCl

CAS RN 135072-15-0

IUPAC Name: 4-benzylmorpholine-2-carboxylic acid hydrochloride
Synonyms: 4-benzylmorpholine-2-carboxylic acid hydrochloride
Molecular Weight (g/mol): 257.71
Molecular Formula: C12H16ClNO3
InChi Key: CEDXMALCJZSQHA-UHFFFAOYNA-N
SMILES: Cl.OC(=O)C1CN(CC2=CC=CC=C2)CCO1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results