CAS RN 135095-52-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
phyproof™ Camelliaside A, ≥90% (HPLC), MilliporeSigma™ Supelco™
This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG