CAS RN 13517-12-9
IUPAC Name:
samarium(3+) hexahydrate tribromide
Synonyms:
samarium(3+) hexahydrate tribromide
Molecular Weight (g/mol):
498.16
Molecular Formula:
Br3H12O6Sm
InChi Key:
LXJDCZVUWJNXRL-UHFFFAOYSA-K
SMILES:
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
1
–
1
of
1
results
1
Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO)
CAS: 13517-12-9 Molecular Formula: Br3H12O6Sm Molecular Weight (g/mol): 498.162 MDL Number: MFCD00192430 InChI Key: LXJDCZVUWJNXRL-UHFFFAOYSA-K Synonym: acmc-20ajrk,samarium bromide hexahydrate,samarium 3+ bromide-water 1/3/6,samarium iii bromide hexahydrate, reacton™ PubChem CID: 57348178 IUPAC Name: samarium(3+);tribromide;hexahydrate SMILES: O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]