accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3C(S)OCH3OCH3CH3NH2CH3

CAS RN 13518-40-6

IUPAC Name: tert-butyl (2S)-2-amino-3-methylbutanoate hydrochloride
Synonyms: tert-butyl (2S)-2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 209.71
Molecular Formula: C9H20ClNO2
InChi Key: AUIVQIHTTVPKFS-FJXQXJEOSA-N
SMILES: Cl.CC(C)[C@H](N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)