Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Valine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. V00975G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00975G Supplier TCI America Supplier No. V00975G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13518-40-6
Molecular Formula C9H20ClNO2
Molecular Weight (g/mol) 209.71
MDL Number MFCD00034863
InChI Key AUIVQIHTTVPKFS-FJXQXJEOSA-N
Synonym h-val-otbu.hcl, l-valine tert-butyl ester hydrochloride, s-tert-butyl 2-amino-3-methylbutanoate hydrochloride, l-valine t-butyl ester hydrochloride, tert-butyl 2s-2-amino-3-methylbutanoate hydrochloride, l-valinetert-butylesterhydrochloride, tert-butyl l-valinate hydrochloride, valine, 1,1-dimethylethyl ester, hydrochloride, h-val-otbu hcl, h-val-obut.hcl
PubChem CID 12490702
IUPAC Name tert-butyl (2S)-2-amino-3-methylbutanoate hydrochloride
SMILES Cl.CC(C)[C@H](N)C(=O)OC(C)(C)C

Specifications

Melting Point 144°C
Color White
Quantity 5 g
Formula Weight 209.71
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Valine tert-Butyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.