accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1351870-16-0

IUPAC Name: 3,6-dibromo-9-(triphenylen-2-yl)-9H-carbazole
Synonyms: 3,6-dibromo-9-(triphenylen-2-yl)carbazole
Molecular Weight (g/mol): 551.28
Molecular Formula: C30H17Br2N
InChi Key: SXVSFEPJEUWLCL-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=C(Br)C=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results