Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromo-9-(triphenylen-2-yl)carbazole 98.0+%, TCI America™
SDP

Catalog No. D4910200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4910200MG 200 mg
D49101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4910200MG Supplier TCI America Supplier No. D4910200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1351870-16-0
Molecular Formula C30H17Br2N
Molecular Weight (g/mol) 551.281
InChI Key SXVSFEPJEUWLCL-UHFFFAOYSA-N
PubChem CID 89492331
IUPAC Name 3,6-dibromo-9-triphenylen-2-ylcarbazole
SMILES C1=CC=C2C(=C1)C3=C(C=C(C=C3)N4C5=C(C=C(C=C5)Br)C6=C4C=CC(=C6)Br)C7=CC=CC=C27

Specifications

Color White
Quantity 200 mg
Formula Weight 551.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(triphenylen-2-yl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.