accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH4CH4CH4CH4CH4CH4NiOClOOOOClOOO

CAS RN 13520-61-1

IUPAC Name: nickel(2+) hexakis(methane) diperchlorate
Synonyms: nickel(2+) hexakis(methane) diperchlorate
Molecular Weight (g/mol): 353.84
Molecular Formula: C6H24Cl2NiO8
InChi Key: GWPPTBKPEOUCMN-UHFFFAOYSA-L
SMILES: C.C.C.C.C.C.[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)