accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OOHUOHNOOOONOO

CAS RN 13520-83-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 504.14
Molecular Formula: H14N2O14U
InChi Key: WRIBVRZWDPGVQH-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O=[U]=O.O[N+]([O-])=O.O[N+]([O-])=O
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)