accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135261-74-4

IUPAC Name: 1-(dimethylamino)-3-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}propan-2-ol hydrochloride
Synonyms: 1-(dimethylamino)-3-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}propan-2-ol hydrochloride
Molecular Weight (g/mol): 365.9
Molecular Formula: C20H28ClNO3
InChi Key: YWZCEYHTWKUNMW-UHFFFAOYNA-N
SMILES: Cl.COC1=CC=CC(CCC2=CC=CC=C2OCC(O)CN(C)C)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results