Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D4355200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4355200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4355200MG Supplier TCI America Supplier No. D4355200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic building block for chemical research.

Chemical Identifiers

CAS 135261-74-4
Molecular Formula C20H28ClNO3
Molecular Weight (g/mol) 365.898
MDL Number MFCD01697013
InChI Key YWZCEYHTWKUNMW-UHFFFAOYSA-N
Synonym 1-(Dimethylamino)-3-[2-[2-(3-methoxyphenyl)ethyl]phenoxy]-2-propanol Hydrochloride
PubChem CID 3070705
IUPAC Name 1-(dimethylamino)-3-[2-[2-(3-methoxyphenyl)ethyl]phenoxy]propan-2-ol;hydrochloride
SMILES CN(C)CC(COC1=CC=CC=C1CCC2=CC(=CC=C2)OC)O.Cl

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 200 mg
Formula Weight 365.90
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1-(Dimethylamino)-3-[2-(3-methoxyphenethyl)phenoxy]-2-propanol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.