CAS RN 1352734-02-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Fluoro-2-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1352734-02-1 Synonym: 4-Fluoro-2-(methoxycarbonylmethyl)phenylboronic acid pinacol ester