accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3ClSiCH3CH3Cl

CAS RN 13528-93-3

IUPAC Name: chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
Synonyms: chloro[2-(chlorodimethylsilyl)ethyl]dimethylsilane
Molecular Weight (g/mol): 215.26
Molecular Formula: C6H16Cl2Si2
InChi Key: VGQOKOYKFDUPPJ-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CC[Si](C)(C)Cl
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (4)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (7)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)