Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorotripyrrolidinophosphonium Hexafluorophosphate

Catalog No. 2656425GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-564-25GF 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 26-564-25GF Supplier Sigma Aldrich Supplier No. 2656425GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for: Peptide coupling reactions and synthesis of coupling reagents Sequence selective peptide recognition Synthesis of condensing reagents Synthesis of heterocyclic +a-sheet ligandsCrystalline reagent for peptide coupling of N-alkylated N-Fmoc and N-Z amino acids in high yield and without racemization.

Specifications

CAS 13528-93-3
Melting Point 35°C to 38°C
Boiling Point 198°C (734 mmHg, lit.)
Molecular Formula C6H16Cl2Si2
Linear Formula [-CH2Si(CH3)2Cl]2
MDL Number MFCD00000505
Quantity 25 g
Molecular Weight (g/mol) 215.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.