accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13534-90-2

IUPAC Name: 3,4-dibromopyridine
Synonyms: 3,4-dibromopyridine
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: ZFZWZGANFCWADD-UHFFFAOYSA-N
SMILES: BrC1=C(Br)C=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results