Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dibromopyridine 97.0+%, TCI America™
SDP

Catalog No. D49161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D49161G Supplier TCI America Supplier No. D49161G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity chemical reagent for use as a heterocyclic building block.

Chemical Identifiers

CAS 13534-90-2
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00234016
InChI Key ZFZWZGANFCWADD-UHFFFAOYSA-N
PubChem CID 817106
IUPAC Name 3,4-dibromopyridine
SMILES C1=CN=CC(=C1Br)Br

Specifications

Melting Point 72°C
Quantity 1 g
Formula Weight 236.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dibromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.