accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2BrN

CAS RN 13534-97-9

IUPAC Name: 6-bromopyridin-3-amine
Synonyms: 6-bromopyridin-3-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: XTHKRYHULUJQHN-UHFFFAOYSA-N
SMILES: NC1=CC=C(Br)N=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)