accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NBr

CAS RN 13535-01-8

IUPAC Name: 5-bromopyridin-3-amine
Synonyms: 5-bromopyridin-3-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: MDQXGHBCDCOOSM-UHFFFAOYSA-N
SMILES: NC1=CN=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results