accessibility menu, dialog, popup

UserName

Viewing profile as user:
1HBr

CAS RN 13536-59-9

IUPAC Name: proton bromide
Synonyms: proton bromide
Molecular Weight (g/mol): 80.91
Molecular Formula: BrH
InChi Key: CPELXLSAUQHCOX-FTGQXOHASA-N
SMILES: [1H+].[Br-]
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results