Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deuterium Bromide 98.0atom%D (45% in D2O), TCI America™
SDP

Catalog No. H03415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H03415G Supplier TCI America Supplier No. H03415G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13536-59-9
Molecular Formula BrH
Molecular Weight (g/mol) 81.918
MDL Number MFCD00044232
InChI Key CPELXLSAUQHCOX-DYCDLGHISA-N
Synonym deuterium bromide, hydrobromic acid-d, deutero-bromide, dbr solution, deuterium bromide solution, bromo 2 h, deuterium bromide, 99 atom % d, deuterium bromide w/w in d2o, deuterium bromide 98.0atom%d in d2o, deuterium bromide solution, 47 wt. % in d2o, 99 atom % d
PubChem CID 518779
SMILES Br

Specifications

UN Number 1788
Quantity 5 g
Formula Weight 81.92
Percent Purity ≥45.0% (T)
Physical Form Liquid
Chemical Name or Material Deuterium Bromide 98.0atom%D (45% in D2O)
EINECSNumber (1)-0105
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.