accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NONH2N

CAS RN 13538-42-6

IUPAC Name: 2-aminopyridine-4-carboxamide
Synonyms: 2-aminopyridine-4-carboxamide
Molecular Weight (g/mol): 137.14
Molecular Formula: C6H7N3O
InChi Key: AGSDASDGMNDAIE-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=NC(N)=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)