Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Amino-isonicotinamide

Catalog No. ADE0001621G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0001621G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0001621G Supplier Sigma Aldrich Supplier No. ADE0001621G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-cyanophenyl sulfofluoridate is an aryl fluorosulfate
  • It undergoes suzuki-miyaura reaction with aryl boronic acid in the presence of pd(oac)2 and et3n to afford the corresponding biaryl derivative
  • 2-cyanophenyl sulfofluoridate can be synthesized from the reaction between phenol and sulfuryl fluoride in the presence of triethylamine.the following aryl fluorosulfate can be utilized as a cross-coupling partner in suzuki-miyaura reactions
  • The standard reaction conditions for these palladium-catalyzed c-c bond formations are ligand-free and can be performed in water, at room temperatre and are not air sensitive.palladium-catalyzed, ligand-free suzuki reaction in water using aryl fluorosulfates

Specifications

CAS 13538-42-6
Molecular Formula C6H7N3O
Linear Formula C6H7N3O
MDL Number MFCD03791262
Quantity 500 mg
Molecular Weight (g/mol) 137.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.