accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNIClN

CAS RN 13544-44-0

IUPAC Name: 2,4-dichloro-5-iodopyrimidine
Synonyms: 2,4-dichloro-5-iodopyrimidine
Molecular Weight (g/mol): 274.87
Molecular Formula: C4HCl2IN2
InChi Key: RGJNPJRAXMSHKN-UHFFFAOYSA-N
SMILES: ClC1=NC=C(I)C(Cl)=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results