Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-iodopyrimidine 98.0+%, TCI America™
SDP

Catalog No. D41225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41225G 5 g
D41221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41225G Supplier TCI America Supplier No. D41225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13544-44-0
Molecular Formula C4HCl2IN2
Molecular Weight (g/mol) 274.87
MDL Number MFCD01898087
InChI Key RGJNPJRAXMSHKN-UHFFFAOYSA-N
PubChem CID 263438
IUPAC Name 2,4-dichloro-5-iodopyrimidine
SMILES C1=C(C(=NC(=N1)Cl)Cl)I

Specifications

Melting Point 70°C
Quantity 5 g
Formula Weight 274.87
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-5-iodopyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.