accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 135447-09-5

IUPAC Name: 2-[(dioctylcarbamoyl)methoxy]acetic acid
Synonyms: [(dioctylcarbamoyl)methoxy]acetic acid
Molecular Weight (g/mol): 357.54
Molecular Formula: C20H39NO4
InChi Key: KOHUSHSNNOEPFN-UHFFFAOYSA-N
SMILES: CCCCCCCCN(CCCCCCCC)C(=O)COCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results