Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Di-n-octyl-3-oxapentanedioic Acid Monoamide 98.0+%, TCI America™
SDP

Catalog No. D43605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D43605G Supplier TCI America Supplier No. D43605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 135447-09-5
Molecular Formula C20H39NO4
Molecular Weight (g/mol) 357.535
InChI Key KOHUSHSNNOEPFN-UHFFFAOYSA-N
Synonym 2-[2-(Di-n-octylamino)-2-oxoethoxy]acetic Acid, DODGAA
PubChem CID 4534750
IUPAC Name 2-[2-(dioctylamino)-2-oxoethoxy]acetic acid
SMILES CCCCCCCCN(CCCCCCCC)C(=O)COCC(=O)O

Specifications

Melting Point 43°C
Color White
Quantity 5 g
Formula Weight 357.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Di-n-octyl-3-oxapentanedioic Acid Monoamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.