accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClNH2H2N

CAS RN 13548-69-1

IUPAC Name: hydrogen 9H-fluorene-2,7-diamine chloride
Synonyms: hydrogen 2,7-diamino-fluoren chloride
Molecular Weight (g/mol): 232.71
Molecular Formula: C13H13ClN2
InChi Key: COOWDVCIUYIBFE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C2C(CC3=CC(N)=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)