Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diaminofluorene Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D00935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00935G 5 g
D009325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D00935G Supplier TCI America Supplier No. D00935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13548-69-1
Molecular Formula C13H14Cl2N2
Molecular Weight (g/mol) 269.169
MDL Number MFCD00012535
InChI Key QJXYCUYLZDQRIN-UHFFFAOYSA-N
Synonym 9h-fluorene-2,7-diamine dihydrochloride, 2,7-diaminofluorene dihydrochloride, 2,7-diamino-fluoren dihydrochloride, 2,7-diaminofluorene di hydrochloride, 2,7-diaminofluorenedihydrochloride, 2,7-diaminofluorene2hcl, 2,7-fluorenyldiamine2hcl, acmc-209c0q, 2,7-diaminofluorene di hcl, 2,7-fluorenediammonium dichloride
PubChem CID 11543576
IUPAC Name 9H-fluorene-2,7-diamine;dihydrochloride
SMILES C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 269.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diaminofluorene Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.