accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2NZnBr

CAS RN 135579-85-0

IUPAC Name: 3-cyanopropan-1-ide; bromozincylium
Synonyms: 3-cyanopropan-1-ide; bromozincylium
Molecular Weight (g/mol): 213.38
Molecular Formula: C4H6BrNZn
InChi Key: HEHCQTGEYNYOSF-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]CCC#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)