Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Ethoxy-4-Oxobutylzinc Bromide Solution

Catalog No. 49849150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-849-150ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 49-849-150ML Supplier Sigma Aldrich Supplier No. 49849150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Ethoxy-4-oxobutylzinc bromide can be used:As an intermediate in one of the key synthetic steps for the preparation of acrylic acid derivatives as EP3 receptor antagonists.As an intermediate in the final step of the total synthesis of a natural product mucosin.As a substrate in the synthesis of multi-substituted haloalkenes by reacting with 1-haloalkenes via Negishi cross-coupling reaction.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 135579-85-0
Density 0.981 g/mL at 25°C
Molecular Formula C4H6BrNZn
Linear Formula NC(CH2)3ZnBr
MDL Number MFCD00671975
Quantity 50 mL
Molecular Weight (g/mol) 213.39 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.