accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2ZnBr

CAS RN 135579-86-1

IUPAC Name: bromozincylium; cyclohexylmethanide
Synonyms: bromozincylium; cyclohexylmethanide
Molecular Weight (g/mol): 242.47
Molecular Formula: C7H13BrZn
InChi Key: SJCALMUTSZRVSS-UHFFFAOYSA-M
SMILES: [Zn+]Br.[CH2-]C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results