Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Cyanobutylzinc Bromide Solution

Catalog No. 49789450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-789-450ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 49-789-450ML Supplier Sigma Aldrich Supplier No. 49789450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Cyanobutylzinc bromide is an organozinc compound, which can be used as a reactant in palladium-catalyzed Negishi cross-coupling reaction to construct carbon-carbon bonds by coupling with organic halides or triflates.It can also be used as a reactant to prepare: 5-(3-Thienyl)pentanenitrile by reacting with 3-bromothiophene in the presence of a nickel catalyst. +a-Cyanobutyl vinylphosphonates by palladium-catalyzed Negishi coupling vinylation reaction with +a-(pseudo)halo vinylphosphonates.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 135579-86-1
Density 0.972 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C7H13BrZn
Linear Formula C6H11CH2ZnBr
MDL Number MFCD01311418
Quantity 50 mL
Molecular Weight (g/mol) 242.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.