accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrFIO

CAS RN 1356113-37-5

IUPAC Name: 3-bromo-4-fluoro-5-iodobenzaldehyde
Synonyms: 3-bromo-4-fluoro-5-iodobenzaldehyde
Molecular Weight (g/mol): 328.91
Molecular Formula: C7H3BrFIO
InChi Key: VZRIOLAMLBZGDB-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=C(C=O)C=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results