accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNBr

CAS RN 13569-96-5

IUPAC Name: 5-(4-bromophenyl)-1H-imidazole
Synonyms: 4-(4-bromophenyl)-3H-imidazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: YLIOAWKNPLJMID-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CN=CN1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)