accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNNNN

CAS RN 1357171-62-0

IUPAC Name: N-({[1,1'-biphenyl]-4-yl}methyl)-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine
Synonyms: N-{[1,1'-biphenyl]-4-ylmethyl}-6-phenyl-3-(pyridin-2-yl)-1,2,4-triazin-5-amine
Molecular Weight (g/mol): 415.5
Molecular Formula: C27H21N5
InChi Key: QNRODODTMXCRKU-UHFFFAOYSA-N
SMILES: C(NC1=C(N=NC(=N1)C1=CC=CC=N1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results