accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13572-98-0

IIIGd

CAS RN 13572-98-0

IUPAC Name: gadolinium(3+) triiodide
Synonyms: gadolinium(3+) triiodide
Molecular Weight (g/mol): 537.96
Molecular Formula: GdI3
InChi Key: IZZTUGMCLUGNPM-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Gd+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results