Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) iodide, ultra dry, 99.99% (REO)

Catalog No. AA3573506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3573506 5 g
AA3573503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3573506 Supplier Thermo Scientific Chemicals Supplier No. 03573506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13572-98-0 | GdI3 | 537.96 g/mol

It is used as a compound in semiconductors and other high purity applications, for contrast imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13572-98-0
Molecular Formula GdI3
Molecular Weight (g/mol) 537.96
MDL Number MFCD00016114
InChI Key IZZTUGMCLUGNPM-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) triiodide
SMILES [I-].[I-].[I-].[Gd+3]

Specifications

Melting Point 926°C
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 537.96
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Gadolinium(III) iodide, Ultra dry
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-997-6
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.