accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3NH3NH4CrSNSNSNSN

CAS RN 13573-16-5

IUPAC Name: chromium(3+) ammonium tetrakis(cyanosulfanide) diamine
Synonyms: reinecke's salt
Molecular Weight (g/mol): 336.41
Molecular Formula: C4H10CrN7S4
InChi Key: ZGLIQORZYPZFPW-UHFFFAOYSA-K
SMILES: N.N.[NH4+].[Cr+3].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
Physical Form 
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Grade 
  • (5)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)