Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reinecke Salt Monohydrate, TCI America™
SDP

Catalog No. R000425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R000425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R000425G Supplier TCI America Supplier No. R000425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory dark purple powder or crystal suitable for use as a synthetic reagent.

Chemical Identifiers

CAS 13573-16-5
Molecular Formula C4H10CrN7S4
Molecular Weight (g/mol) 354.43
MDL Number MFCD00150925

Specifications

Melting Point 270°C
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 336.41
Chemical Name or Material Reinecke Salt Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.