accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13574-13-5

IUPAC Name: (2S)-5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
Synonyms: (2S)-5-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoate
Molecular Weight (g/mol): 336.37
Molecular Formula: C17H22NO6
InChi Key: AJDUMMXHVCMISJ-ZDUSSCGKSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)