Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate 98.0+%, TCI America™
SDP

Catalog No. B16335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16335G 5 g
B163325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16335G Supplier TCI America Supplier No. B16335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13574-13-5
Molecular Formula C17H22NO6
Molecular Weight (g/mol) 336.37
MDL Number MFCD00065569
InChI Key AJDUMMXHVCMISJ-ZDUSSCGKSA-M
Synonym boc-glu obzl-oh, boc-l-glutamic acid 5-benzyl ester, boc-l-glutamic acid 5-benzylester, 5-benzyl n-boc-l-glutamate, boc-l-glu obzl-oh, n-alpha-tert-boc-l-glutamic-gamma-benzyl ester, 5-benzyl n-tert-butoxycarbonyl-l-glutamate, boc-l-glutamic acid-5-benzyl ester, l-glutamic acid, n-1,1-dimethylethoxy carbonyl-, 5-phenylmethyl ester, n-boc-l-glutamic acid 5-benzyl ester
PubChem CID 83589
IUPAC Name (2S)-5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
SMILES CC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O

Specifications

Melting Point 68°C
Color White
Quantity 5 g
Formula Weight 337.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Benzyl N-(tert-Butoxycarbonyl)-L-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.