accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HOHNNH2HClHCl

CAS RN 1357471-44-3

IUPAC Name: N1-(naphthalen-1-yl)ethane-1,2-diamine methanol dihydrochloride
Synonyms: N-(1-naphthyl)ethylenediamine methanol dihydrochloride
Molecular Weight (g/mol): 291.22
Molecular Formula: C13H20Cl2N2O
InChi Key: YUMCJWVZLMGNTD-UHFFFAOYSA-N
SMILES: Cl.Cl.CO.NCCNC1=CC=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results