Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(1-Naphthyl)ethylenediamine Dihydrochloride Monomethanolate 97.0+%, TCI America™
SDP

Catalog No. N057725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N057725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N057725G Supplier TCI America Supplier No. N057725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1357471-44-3
Molecular Formula C13H20Cl2N2O
Molecular Weight (g/mol) 291.22
MDL Number MFCD00012556
InChI Key YUMCJWVZLMGNTD-UHFFFAOYSA-N
Synonym marshall's reagent, n-1-naphthylethylenediamine dihydrochloride monomethanolate, n-1-naphthyl ethylenediamine dihydrochloride monomethanolate, n-1-naphthyl ethylenediamine dihydrochloride monomethanolate, for spectrophotometric det. of nitrate and nitrite
PubChem CID 44630407

Specifications

Melting Point 190°C
Quantity 25 g
Formula Weight 291.22
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(1-Naphthyl)ethylenediamine Dihydrochloride Monomethanolate
EINECSNumber (9)-0935
RTECSNumber KV5330000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.